missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC433890050

 View more versions of this product

Catalog No. AC433890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Nitrophenylboronic acid
Glass bottle
5g
96.0
97%
MFCD00161360
NSFJAFZHYOAMHL-UHFFFAOYSA-N
(4-nitrophenyl)boronic acid
2773552
Crystalline Powder
Authentic
White to Yellow
24067-17-2
100.0
C6H6BNO4
4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
B(C1=CC=C(C=C1)[N+](=O)[O-])(O)O
166.927
166.93
≥96.0%

Chemical Identifiers

24067-17-2
166.927
NSFJAFZHYOAMHL-UHFFFAOYSA-N
2773552
B(C1=CC=C(C=C1)[N+](=O)[O-])(O)O
C6H6BNO4
MFCD00161360
4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
(4-nitrophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh and keep confortable for several minutes.

Warning

SDS Documents