missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenylacetonitrile, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC166065000

 View more versions of this product

Catalog No. AC166065000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Nitrophenylacetonitrile
Glass bottle
Cream to Yellow
500g
97.5
98%
O2NC6H4CH2CN
09, 456
2-4-nitrophenyl acetonitrile
PXNJGLAVKOXITN-UHFFFAOYSA-N
2-(4-nitrophenyl)acetonitrile
68386
Crystalline Powder
Authentic
195°C to 197°C (16.0 hPa)
114°C to 118°C
555-21-5
100.0
C8H6N2O2
MFCD00007372
15, 6678
Solubility in water: insoluble. Other solubilities: soluble in alchol,ether and benzene
C1=CC(=CC=C1CC#N)[N+](=O)[O-]
162.15
162.15
98%

Chemical Identifiers

555-21-5
162.15
PXNJGLAVKOXITN-UHFFFAOYSA-N
68386
C1=CC(=CC=C1CC#N)[N+](=O)[O-]
C8H6N2O2
MFCD00007372
2-4-nitrophenyl acetonitrile
2-(4-nitrophenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-085-

SDS Documents