Learn More
4-Nitrophenylacetonitrile, 98%, ACROS Organics™
Chemical Identifiers
CAS | 555-21-5 |
---|---|
Molecular Formula | C8H6N2O2 |
Molecular Weight (g/mol) | 162.15 |
MDL Number | MFCD00007372 |
InChI Key | PXNJGLAVKOXITN-UHFFFAOYSA-N |
Synonym | 2-4-nitrophenyl acetonitrile, 4-nitrobenzeneacetonitrile, 4-nitrobenzyl cyanide, 4-nitrophenylacetonitrile, acetonitrile, p-nitrophenyl, benzeneacetonitrile, 4-nitro, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, p-nitrophenyl acetonitrile, p-nitrophenylacetonitrile |
PubChem CID | 68386 |
IUPAC Name | 2-(4-nitrophenyl)acetonitrile |
SMILES | C1=CC(=CC=C1CC#N)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC166060250
|
Thermo Fisher Scientific
166060250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC166061000
|
Thermo Fisher Scientific
166061000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC166065000
|
Thermo Fisher Scientific
AC166065000 |
500g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
555-21-5 | |
162.15 | |
PXNJGLAVKOXITN-UHFFFAOYSA-N | |
68386 | |
C1=CC(=CC=C1CC#N)[N+](=O)[O-] |
C8H6N2O2 | |
MFCD00007372 | |
2-4-nitrophenyl acetonitrile, 4-nitrobenzeneacetonitrile, 4-nitrobenzyl cyanide, 4-nitrophenylacetonitrile, acetonitrile, p-nitrophenyl, benzeneacetonitrile, 4-nitro, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, p-nitrophenyl acetonitrile, p-nitrophenylacetonitrile | |
2-(4-nitrophenyl)acetonitrile |
Specifications
Authentic | |
195°C to 197°C (16.0 hPa) | |
114°C to 118°C | |
555-21-5 | |
100.0 | |
C8H6N2O2 | |
MFCD00007372 | |
15, 6678 | |
Solubility in water: insoluble. Other solubilities: soluble in alchol,ether and benzene | |
C1=CC(=CC=C1CC#N)[N+](=O)[O-] | |
162.15 | |
162.15 | |
98% |
Glass bottle | |
Cream to Yellow | |
25g | |
97.5 | |
98% | |
O2NC6H4CH2CN | |
09, 456 | |
2-4-nitrophenyl acetonitrile, 4-nitrobenzeneacetonitrile, 4-nitrobenzyl cyanide, 4-nitrophenylacetonitrile, acetonitrile, p-nitrophenyl, benzeneacetonitrile, 4-nitro, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, p-nitrophenyl acetonitrile, p-nitrophenylacetonitrile | |
PXNJGLAVKOXITN-UHFFFAOYSA-N | |
2-(4-nitrophenyl)acetonitrile | |
68386 | |
Crystalline Powder | |
4-Nitrophenylacetonitrile |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-085-