missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenylacetonitrile, 98%, ACROS Organics™

Manufacturer:  Acros Organics 166060250

 View more versions of this product

Catalog No. AC166060250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Nitrophenylacetonitrile
Glass bottle
114°C to 118°C
555-21-5
100.0
C8H6N2O2
MFCD00007372
15, 6678
PXNJGLAVKOXITN-UHFFFAOYSA-N
2-(4-nitrophenyl)acetonitrile
68386
Crystalline Powder
Authentic
Cream to Yellow
25g
97.5
98%
O2NC6H4CH2CN
09, 456
4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
C1=CC(=CC=C1CC#N)[N+](=O)[O-]
162.148
162.15
≥97.5%

Chemical Identifiers

555-21-5
162.148
PXNJGLAVKOXITN-UHFFFAOYSA-N
68386
C1=CC(=CC=C1CC#N)[N+](=O)[O-]
C8H6N2O2
MFCD00007372
4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
2-(4-nitrophenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents