missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl Trifluoromethanesulfonate, TCI America™

$36.67 - $123.75

Chemical Identifiers

CAS 17763-80-3
Molecular Formula C7H4F3NO5S
Molecular Weight (g/mol) 271.166
MDL Number MFCD00192354
InChI Key NDTIXHNCNLKURN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 4-nitrophenyl trifluoromethanesulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
PubChem CID 549870
IUPAC Name (4-nitrophenyl) trifluoromethanesulfonate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N099325G View Documents TCI America
N099325G
25g Each for $123.75
 
N09935G View Documents TCI America
N09935G
5g Each for $36.67
 
Specifications

Chemical Identifiers

17763-80-3
271.166
NDTIXHNCNLKURN-UHFFFAOYSA-N
549870
C1=CC(=CC=C1[N+](=O)[O-])OS(=O)(=O)C(F)(F)F
C7H4F3NO5S
MFCD00192354
trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 4-nitrophenyl trifluoromethanesulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
(4-nitrophenyl) trifluoromethanesulfonate

Specifications

Yellow
25g
C7H4F3NO5S
3261
NDTIXHNCNLKURN-UHFFFAOYSA-N
(4-nitrophenyl) trifluoromethanesulfonate
549870
Crystal-Powder at 20°C
4-Nitrophenyl Trifluoromethanesulfonate
54°C
17763-80-3
MFCD00192354
trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 4-nitrophenyl trifluoromethanesulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])OS(=O)(=O)C(F)(F)F
271.166
271.17
≥99.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No