missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC128860500

 View more versions of this product

Catalog No. AC128860500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Nitrophenyl phosphate, disodium salt, hexahydrate
4264-83-9
Glass bottle
≥98.0%
C6H4NNa2O6P·6H2O
White to slightly yellow
50g
(5% aq. soln.) clear
Authentic
Crystalline Powder or Crystals
98+%
O2NC6H4OP(O)(ONa)2·6H2O
371.13
PNPP

Chemical Identifiers

98+%
O2NC6H4OP(O)(ONa)2·6H2O
PNPP
≥98.0%
C6H4NNa2O6P·6H2O
371.13
4-Nitrophenyl phosphate, disodium salt, hexahydrate