Please login to your online account to display your discounted pricing

4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate, 99+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC227990250

 View more versions of this product

Catalog No. AC227990250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder
Formula Weight 435.47
Quantity 25g
Color White to slightly yellow
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate
Packaging Glass bottle
Name Note 99+%
Water 5% max.
Assay Percent Range 99+%
Molecular Formula C18H32N3O6P·H2O
Linear Formula O2NC6H4O3P(O)(H3NC6H11)2·H2O
MDL Number MFCD00066287
CAS 123334-11-2
Synonym 4-Nitrophenyl phosphate di(cyclohexylamine) salt monohydrate
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate
Name Note 99+%
CAS 123334-11-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C18H32N3O6P·H2O
Linear Formula O2NC6H4O3P(O)(H3NC6H11)2·H2O
Formula Weight 435.47
MDL Number MFCD00066287
Synonym 4-Nitrophenyl phosphate di(cyclohexylamine) salt monohydrate
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents