Please login to your online account to display your discounted pricing

4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate, 99+%, ACROS Organics™

$30.53 - $106.28

Chemical Identifiers

Chemical Name or Material 4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate
Name Note 99+%
CAS 123334-11-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C18H32N3O6P·H2O
Linear Formula O2NC6H4O3P(O)(H3NC6H11)2·H2O
Formula Weight 435.47
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC227990250

  SDS
thermo fisher scientific
AC227990250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC227990010

  SDS
thermo fisher scientific
AC227990010
Glass bottle 1g Each for $30.53

AC227990050

  SDS
thermo fisher scientific
227990050
Glass bottle 5g Each for $106.28
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 435.47
Physical Form Powder
Quantity 25g
Color White to slightly yellow
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 4-Nitrophenyl phosphate di(cyclohexylamine) salt monohydrate
Water 5% max.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Name Note 99+%
Chemical Name or Material 4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate
Molecular Formula C18H32N3O6P·H2O
MDL Number MFCD00066287
CAS 123334-11-2
Assay Percent Range 99+%
Linear Formula O2NC6H4O3P(O)(H3NC6H11)2·H2O

Phase transfer catalyst

Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.