Please login to your online account to display your discounted pricing

4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC227990010

 View more versions of this product

Catalog No. AC227990010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 435.47
Physical Form Powder
Quantity 1g
Color White to slightly yellow
CAS Min % 99.0
CAS Max % 100.0
Water 5% max.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym 4-Nitrophenyl phosphate di(cyclohexylamine) salt monohydrate
Linear Formula O2NC6H4O3P(O)(H3NC6H11)2·H2O
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Assay Percent Range 99+%
Chemical Name or Material 4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate
Name Note 99+%
MDL Number MFCD00066287
Molecular Formula C18H32N3O6P·H2O
CAS 123334-11-2

Specifications

Chemical Name or Material 4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt monohydrate
Name Note 99+%
CAS 123334-11-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C18H32N3O6P·H2O
Linear Formula O2NC6H4O3P(O)(H3NC6H11)2·H2O
Formula Weight 435.47
MDL Number MFCD00066287
Synonym 4-Nitrophenyl phosphate di(cyclohexylamine) salt monohydrate
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents