missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl phenyl sulfide, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 181170250

 View more versions of this product

Catalog No. AC181170250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
231.27
Yellow
54° to 58°C
97.5
Glass bottle
4-Nitrophenyl phenyl sulfide
C12 H9 N O2 S
MFCD00024700
Crystalline Powder
25g
06, 339
100.0
Authentic
952-97-6
O2NC6H4SC6H5
98%
98%

Chemical Identifiers

952-97-6
100.0
C12 H9 N O2 S
231.27
4-Nitrophenyl phenyl sulfide, 98%
97.5
98%
O2NC6H4SC6H5
MFCD00024700
≥97.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 213-462-5

RTECSNumber : WQ5620000