Please login to your online account to display your discounted pricing

4-Nitrophenyl phenyl sulfide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  181170050

 View more versions of this product

Catalog No. AC181170050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 231.27
Quantity 5g
Color Yellow
EINECS Number 213-462-5
RTECS Number WQ5620000
Melting Point 54° to 58°C
Beilstein 06, 339
CAS Max % 100.0
Chemical Name or Material 4-Nitrophenyl phenyl sulfide
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C12 H9 N O2 S
Linear Formula O2NC6H4SC6H5
MDL Number MFCD00024700
CAS 952-97-6
CAS Min % 97.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Nitrophenyl phenyl sulfide
Name Note 98%
CAS 952-97-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12 H9 N O2 S
Linear Formula O2NC6H4SC6H5
Formula Weight 231.27
MDL Number MFCD00024700