missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Nitrophenyl)(phenyl)iodonium Trifluoromethanesulfonate, TCI America™

$317.78 - $1,109.19

Chemical Identifiers

CAS 905718-45-8
Molecular Formula C13H9F3INO5S
Molecular Weight (g/mol) 475.177
MDL Number MFCD21608480
InChI Key OPSOSRFTIUMCJF-UHFFFAOYSA-M
Synonym (4-Nitrophenyl)(phenyl)iodonium Triflate
PubChem CID 16727017
IUPAC Name (4-nitrophenyl)-phenyliodanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[I+]C2=CC=C(C=C2)[N+](=O)[O-].C(F)(F)(F)S(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10661G View Documents TCI America
N10661G
1g Each for $317.78
 
N10665G View Documents TCI America
N10665G
5g Each for $1,109.19
 
Specifications

Chemical Identifiers

905718-45-8
475.177
OPSOSRFTIUMCJF-UHFFFAOYSA-M
16727017
C1=CC=C(C=C1)[I+]C2=CC=C(C=C2)[N+](=O)[O-].C(F)(F)(F)S(=O)(=O)[O-]
C13H9F3INO5S
MFCD21608480
(4-Nitrophenyl)(phenyl)iodonium Triflate
(4-nitrophenyl)-phenyliodanium;trifluoromethanesulfonate

Specifications

Undesignated
1g
C13H9F3INO5S
(4-Nitrophenyl)(phenyl)iodonium Triflate
C1=CC=C(C=C1)[I+]C2=CC=C(C=C2)[N+](=O)[O-].C(F)(F)(F)S(=O)(=O)[O-]
475.177
475.18
≥98.0% (HPLC)
200°C
905718-45-8
MFCD21608480
OPSOSRFTIUMCJF-UHFFFAOYSA-M
(4-nitrophenyl)-phenyliodanium;trifluoromethanesulfonate
16727017
Crystal-Powder at 20°C
(4-Nitrophenyl)(phenyl)iodonium Trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No