Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenyl phenyl ether, 99%

$30.58 - $165.74

Chemical Identifiers

Chemical Name or Material 4-Nitrophenyl phenyl ether
CAS 620-88-2
Molecular Formula C12H9NO3
Formula Weight 215.21
MDL Number MFCD00007320
Synonym p-Nitrodiphenylether; 1-Nitro-4-phenoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2559606

  SDS
alfa aesar™
B2559606
5g Each for $30.58

AAB2559614

  SDS
alfa aesar™
B2559614
25g Each for $90.41

AAB2559622

  SDS
alfa aesar™
B2559622
100g Each for $165.74
Description & Specifications

Specifications

Formula Weight 215.21
Chemical Name or Material 4-Nitrophenyl phenyl ether
Boiling Point 320°C
Molecular Formula C12H9NO3
MDL Number MFCD00007320
CAS 620-88-2
Synonym p-Nitrodiphenylether; 1-Nitro-4-phenoxybenzene
RTECS Number KO2680000
Flash Point >110°C (230°F)
Melting Point 53° to 56°C
TSCA TSCA
Quantity 5g
Assay 99%