Please login to your online account to display your discounted pricing

4-Nitrophenyl octyl ether, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  33748-0050

 View more versions of this product

Catalog No. AC337480050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid After Melting
Formula Weight 251.32
Quantity 5g
Color Clear yellow
CAS Max % 100.0
Infrared Spectrum Authentic
Storage Information Product may darken during storage
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Linear Formula NO2C6H4O(CH2)7CH3
MDL Number MFCD00043611
Chemical Name or Material 4-Nitrophenyl octyl ether
CAS 49562-76-7
Molecular Formula C14 H21 N O3
Assay Percent Range 99%

Specifications

Chemical Name or Material 4-Nitrophenyl octyl ether
Name Note 99%
CAS 49562-76-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14 H21 N O3
Linear Formula NO2C6H4O(CH2)7CH3
Formula Weight 251.32
MDL Number MFCD00043611
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents