Please login to your online account to display your discounted pricing

4-Nitrophenyl octyl ether, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Nitrophenyl octyl ether
Name Note 99%
CAS 49562-76-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14 H21 N O3
Linear Formula NO2C6H4O(CH2)7CH3
Formula Weight 251.32
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC337480050

  SDS
thermo fisher scientific
33748-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid After Melting
Formula Weight 251.32
Quantity 5g
Color Clear yellow
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 4-Nitrophenyl octyl ether
Packaging Glass bottle
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Storage Information Product may darken during storage
Name Note 99%
Assay Percent Range 99%
Molecular Formula C14 H21 N O3
Linear Formula NO2C6H4O(CH2)7CH3
MDL Number MFCD00043611
CAS 49562-76-7
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.