missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl isothiocyanate, 97%, ACROS Organics™

Chemical Identifiers

CAS 2131-61-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H4N2O2S
Linear Formula NCSC6H4NO2
Formula Weight 180.19
Chemical Name or Material 4-Nitrophenyl isothiocyanate
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC416070250 SDS Thermo Fisher Scientific
416070250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2131-61-5
100.0
C7H4N2O2S
180.19
≥97.5%
97.5
98%
NCSC6H4NO2
4-Nitrophenyl isothiocyanate

Specifications

180.19
25g
109° to 113°C
Authentic
97.5
NCSC6H4NO2
98%
≥97.5%
Crystalline Powder
Yellow
100.0
C7H4N2O2S
Glass bottle
2131-61-5
4-Nitrophenyl isothiocyanate