missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl Isocyanide, TCI America™

$177.22 - $621.81

Chemical Identifiers

CAS 1984-23-2
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.121
MDL Number MFCD01937394
InChI Key WQFPYEBGLBCQOY-UHFFFAOYSA-N
Synonym 1-Isocyano-4-nitrobenzene
PubChem CID 74802
IUPAC Name 1-isocyano-4-nitrobenzene
SMILES [C-]#[N+]C1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09821G View Documents TCI America
N09821G
1g Each for $621.81
 
N0982200MG View Documents TCI America
N0982200MG
200mg Each for $177.22
 
Specifications

Chemical Identifiers

1984-23-2
148.121
WQFPYEBGLBCQOY-UHFFFAOYSA-N
74802
[C-]#[N+]C1=CC=C(C=C1)[N+](=O)[O-]
C7H4N2O2
MFCD01937394
1-Isocyano-4-nitrobenzene
1-isocyano-4-nitrobenzene

Specifications

Yellow
1g
C7H4N2O2
1-Isocyano-4-nitrobenzene
[C-]#[N+]C1=CC=C(C=C1)[N+](=O)[O-]
148.121
148.12
≥98.0% (GC)
115°C
1984-23-2
MFCD01937394
WQFPYEBGLBCQOY-UHFFFAOYSA-N
1-isocyano-4-nitrobenzene
74802
Crystal-Powder at 20°C
4-Nitrophenyl Isocyanide
Safety and Handling

Safety and Handling

RTECSNumber : DA3768224

TSCA : No