missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl Isocyanide 98.0+%, TCI America™

Manufacturer:  TCI America N0982200MG

 View more versions of this product

Catalog No. N0982200MG



Specifications

Specifications

4-Nitrophenyl Isocyanide
115°C
1984-23-2
MFCD01937394
WQFPYEBGLBCQOY-UHFFFAOYSA-N
1-isocyano-4-nitrobenzene
74802
Crystal-Powder at 20°C
Yellow
200mg
C7H4N2O2
1-Isocyano-4-nitrobenzene
[C-]#[N+]C1=CC=C(C=C1)[N+](=O)[O-]
148.121
148.12
≥98.0% (GC)

Chemical Identifiers

1984-23-2
148.121
WQFPYEBGLBCQOY-UHFFFAOYSA-N
74802
[C-]#[N+]C1=CC=C(C=C1)[N+](=O)[O-]
C7H4N2O2
MFCD01937394
1-Isocyano-4-nitrobenzene
1-isocyano-4-nitrobenzene
Safety and Handling

Safety and Handling

RTECSNumber : DA3768224

TSCA : No

Documents