missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenyl isocyanate, 97%

Manufacturer:  Alfa Aesar™ A1119403

 View more versions of this product

Catalog No. AAA1119403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.
Specifications

Specifications

4-Nitrophenyl isocyanate
137°C to 138°C (11mmHg)
1g
C7H4N2O3
UN3335
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
C1=CC(=CC=C1N=C=O)[N+](=O)[O-]
164.12
164.12
>110°C (230°F)
56°C to 59°C
100-28-7
MFCD00007306
389516
GFNKTLQTQSALEJ-UHFFFAOYSA-N
1-isocyanato-4-nitrobenzene
66012
97%

Chemical Identifiers

100-28-7
164.12
GFNKTLQTQSALEJ-UHFFFAOYSA-N
66012
C1=CC(=CC=C1N=C=O)[N+](=O)[O-]
C7H4N2O3
MFCD00007306
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
1-isocyanato-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 9; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.

RTECSNumber : DA3692500

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents