Please login to your online account to display your discounted pricing

4-Nitrophenyl formate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Nitrophenyl formate
CAS 1865-01-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H5NO4
Formula Weight 167.12
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC438780050

 
thermo fisher scientific
AC438780050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Crystals
Formula Weight 167.12
Quantity 5g
Color White to off-white
Melting Point 66.0° to 73.0°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H5NO4
CAS 1865-01-6
Chemical Name or Material 4-Nitrophenyl formate
Elemental Analysis 48.30 to 52.32% C, 8.05 to 8.72% N