missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl formate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438780050

 View more versions of this product

Catalog No. AC438780050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Nitrophenyl formate
Authentic
White
5g
96.0
97%
4-nitrophenyl formate, 4-nitrophenyl formate hplc, acmc-1bty1, formic acid 4-nitro-phenyl ester, formic acid 4-nitrophenyl ester, formic acid, 4-nitrophenyl ester, formic acid, p-nitrophenyl ester, formic acid,4-nitrophenyl ester, p-nitrophenyl formate
C1=CC(=CC=C1[N+](=O)[O-])OC=O
167.12
167.12
97%
48.30 to 52.32% C, 8.05 to 8.72% N
Glass bottle
66.0°C to 67.0°C
1865-01-6
100.0
C7H5NO4
IEXRKQFZXJSHOB-UHFFFAOYSA-N
(4-nitrophenyl) formate
74628
Crystalline Powder or Crystals

Chemical Identifiers

1865-01-6
167.12
4-nitrophenyl formate, 4-nitrophenyl formate hplc, acmc-1bty1, formic acid 4-nitro-phenyl ester, formic acid 4-nitrophenyl ester, formic acid, 4-nitrophenyl ester, formic acid, p-nitrophenyl ester, formic acid,4-nitrophenyl ester, p-nitrophenyl formate
(4-nitrophenyl) formate
C7H5NO4
IEXRKQFZXJSHOB-UHFFFAOYSA-N
74628
C1=CC(=CC=C1[N+](=O)[O-])OC=O
Safety and Handling

Safety and Handling

EINECSNumber : 217-473-6

SDS Documents