missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl-β-D-fucopyranoside, ACROS Organics™

$54.00 - $54.00

Chemical Identifiers

CAS 1226-39-7
Molecular Formula C12 H15 N O7
Formula Weight 285.25
MDL Number MFCD00040651
Synonym p-Nitrophenyl-α-D-fucoside
Chemical Name or Material 4-Nitrophenyl-β-D-fucopyranoside
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC336850250 SDS Thermo Fisher Scientific
AC336850250
Glass bottle 25mg Each for $54.00
Specifications Description & Specifications

Chemical Identifiers

1226-39-7
285.25
p-Nitrophenyl-α-D-fucoside
C12 H15 N O7
MFCD00040651
4-Nitrophenyl-β-D-fucopyranoside

Specifications

1226-39-7
C12 H15 N O7
MFCD00040651
p-Nitrophenyl-α-D-fucoside
Glass bottle
285.25
25mg
4-Nitrophenyl-β-D-fucopyranoside