missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl chloroformate, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC170805000

 View more versions of this product

Catalog No. AC170805000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Coupling reagent
Specifications

Specifications

4-Nitrophenyl chloroformate
Authentic
159.0°C to 162.0°C (19.0mmHg)
73.0°C to 81.0°C
7693-46-1
100.0
C7H4ClNO4
MFCD00007321
02,297
Solubility in water: decomposes. Other solubilities: soluble in toluene and benzene
C1=CC(=CC=C1[N+](=O)[O-])OC(=O)Cl
201.562
201.57
97%
>110°C
Glass bottle
Beige-Yellow to White
500g
96.0
97%
ClCO2C6H4NO2
06, I, 120
4-nitrophenyl chloroformate, carbonochloridic acid, 4-nitrophenyl ester, chloroformic acid 4-nitrophenyl ester, p-nitrophenyl chloroformate, chloroformic acid p-nitrophenyl ester, 4-nitrophenyl carbonochloridate, 4-nitrophenyl chloroformate, formic acid, chloro-, p-nitrophenyl ester, 4-nitrophenylchlorocarbonat, 4-nitrophenyl choroformate
NXLNNXIXOYSCMB-UHFFFAOYSA-N
(4-nitrophenyl) carbonochloridate
82129
Crystalline Powder, Crystals and/or Chunks

Chemical Identifiers

7693-46-1
201.562
NXLNNXIXOYSCMB-UHFFFAOYSA-N
82129
C1=CC(=CC=C1[N+](=O)[O-])OC(=O)Cl
C7H4ClNO4
MFCD00007321
4-nitrophenyl chloroformate, carbonochloridic acid, 4-nitrophenyl ester, chloroformic acid 4-nitrophenyl ester, p-nitrophenyl chloroformate, chloroformic acid p-nitrophenyl ester, 4-nitrophenyl carbonochloridate, 4-nitrophenyl chloroformate, formic acid, chloro-, p-nitrophenyl ester, 4-nitrophenylchlorocarbonat, 4-nitrophenyl choroformate
(4-nitrophenyl) carbonochloridate
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed. Toxic if inhaled. May cause respiratory irritation. Toxic in contact with skin. Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

SDS Documents