Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenyl bromoacetate, 98%

$70.35 - $238.59

Chemical Identifiers

Chemical Name or Material 4-Nitrophenyl bromoacetate
CAS 19199-82-7
Molecular Formula C8H6BrNO4
Formula Weight 260.05
MDL Number MFCD00051840
Beilstein 2529186
Synonym Bromoacetic acid p-nitrophenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0800009

 
alfa aesar™
L0800009
10g Each for $70.35

AAL0800018

 
alfa aesar™
L0800018
50g Each for $238.59
Description & Specifications

Specifications

Formula Weight 260.05
Chemical Name or Material 4-Nitrophenyl bromoacetate
MDL Number MFCD00051840
Assay 98%
Molecular Formula C8H6BrNO4
Melting Point 84° to 88°C
CAS 19199-82-7
Quantity 10g
Synonym Bromoacetic acid p-nitrophenyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Beilstein 2529186

Applications
4-Nitrophenyl bromoacetate is used as a haloacetamide

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

2° to 8°C