Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenyl acetate, 98+%


Quantity 50g

Manufacturer: alfa aesar™  L0031418

 View more versions of this product

Catalog No. AAL0031418

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 181.15
Chemical Name or Material 4-Nitrophenyl acetate
CAS 830-03-5
Molecular Formula C8H7NO4
Synonym Acetic acid 4-nitrophenyl ester
TSCA TSCA
Melting Point 77° to 79°C
Beilstein 515874
RTECS Number AJ1150000
Quantity 50g
Assay ≥98%
MDL Number MFCD00007326

Specifications

Chemical Name or Material 4-Nitrophenyl acetate
CAS 830-03-5
Molecular Formula C8H7NO4
Formula Weight 181.15
MDL Number MFCD00007326
Beilstein 515874
Synonym Acetic acid 4-nitrophenyl ester

Applications
4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.