Please login to your online account to display your discounted pricing

4-Nitrophenyl 2-(trimethylsilyl)ethylecarbonate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Nitrophenyl 2-(trimethylsilyl)ethylecarbonate
CAS 80149-80-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C12H17NO5Si
Formula Weight 283.36
Synonym 2-(Trimethylsilyl)ethyle4-nitrophenyl carbonate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC440070050

  SDS
thermo fisher scientific
AC440070050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440070250

  SDS
thermo fisher scientific
AC440070250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥94.0%
Formula Weight 283.36
Quantity 5g
Color Pale yellow
Melting Point 35° to 40°C
CAS Min % 94.0
CAS Max % 100.0
Synonym 2-(Trimethylsilyl)ethyle4-nitrophenyl carbonate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Chemical Name or Material 4-Nitrophenyl 2-(trimethylsilyl)ethylecarbonate
Assay Percent Range 95%
Molecular Formula C12H17NO5Si
CAS 80149-80-0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.