missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate, 95%

$78.03 - $306.88

Chemical Identifiers

CAS 80149-80-0
Molecular Formula C12H17NO5Si
Molecular Weight (g/mol) 283.355
MDL Number MFCD00042930
InChI Key ZAQWGGKIMQIVGM-UHFFFAOYSA-N
Synonym 4-nitrophenyl 2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204
PubChem CID 3086116
IUPAC Name (4-nitrophenyl) 2-trimethylsilylethyl carbonate
SMILES C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6046603 View Documents Alfa Aesar™
H6046603
1g Each for $78.03
 
AAH6046606 View Documents Alfa Aesar™
H6046606
5g Each for $306.88
 
Description

Description

Applications
It is employed as a reagent for the introduction of the TEOC-amino protecting group which can be cleaved by F-.

Solubility
Slightly soluble in water.

Notes
Moisture & Light Sensitive. Store at 2-8°C.
Specifications

Chemical Identifiers

80149-80-0
283.355
ZAQWGGKIMQIVGM-UHFFFAOYSA-N
3086116
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
C12H17NO5Si
MFCD00042930
4-nitrophenyl 2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204
(4-nitrophenyl) 2-trimethylsilylethyl carbonate

Specifications

35°C to 40°C
80149-80-0
C12H17NO5Si
4320585
ZAQWGGKIMQIVGM-UHFFFAOYSA-N
(4-nitrophenyl) 2-trimethylsilylethyl carbonate
3086116
95%
1g
C12H17NO5Si
MFCD00042930
4-nitrophenyl 2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
283.355
283.35
4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C