missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC229410010

 View more versions of this product

Catalog No. AC229410010



Specifications

Specifications

(0.5% aq. soln.) clear, colorless
Authentic
−16° (20°C c=0.5,water)
0.5% max. (K.F.)
210.0°C to 212.0°C
99%
98.5
98.5% min. (HPLC)
MFCD00063696
4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4-nitrophenyl-n-acetyl-beta-d-glucosaminide, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, p-nitrophenyl n-acetyl-beta-d-glucosaminide
OMRLTNCLYHKQCK-DHGKCCLASA-N
N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
102416
Fine Powder
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside
Glass bottle
− 16.00
White
1g
3459-18-5
100.0
C14H18N2O8
18, V,11, 102
(0.5% in water) Clear colorless to light yellow
CC(=O)NC1C(C(C(OC1OC2=CC=C(C=C2)[N+](=O)[O-])CO)O)O
342.29
342.29
99%

Chemical Identifiers

3459-18-5
342.29
OMRLTNCLYHKQCK-DHGKCCLASA-N
102416
CC(=O)NC1C(C(C(OC1OC2=CC=C(C=C2)[N+](=O)[O-])CO)O)O
C14H18N2O8
MFCD00063696
4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4-nitrophenyl-n-acetyl-beta-d-glucosaminide, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, p-nitrophenyl n-acetyl-beta-d-glucosaminide
N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
Safety and Handling

Safety and Handling

EINECSNumber : 222-398-7

SDS Documents