missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenoxyacetic Acid, TCI America™

$85.56 - $550.02

Chemical Identifiers

CAS 1798-11-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00017030
InChI Key AVDLFIONKHGQAP-UHFFFAOYSA-N
Synonym 4-nitrophenoxyacetic acid, 2-4-nitrophenoxy acetic acid, 4-nitrophenoxy acetic acid, p-nitrophenoxyacetic acid, acetic acid, 4-nitrophenoxy, 4-nitro-phenoxy-acetic acid, acetic acid, p-nitrophenoxy, acetic acid, 2-4-nitrophenoxy, acmc-209efx, 4-nitrophenoxy acetic acid
PubChem CID 15720
SMILES C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N022625G View Documents TCI America
N022625G
25g Each for $85.56
 
N0226500G View Documents TCI America
N0226500G
500g Each for $550.02
 
Specifications

Chemical Identifiers

1798-11-4
197.146
AVDLFIONKHGQAP-UHFFFAOYSA-N
15720
C8H7NO5
MFCD00017030
4-nitrophenoxyacetic acid, 2-4-nitrophenoxy acetic acid, 4-nitrophenoxy acetic acid, p-nitrophenoxyacetic acid, acetic acid, 4-nitrophenoxy, 4-nitro-phenoxy-acetic acid, acetic acid, p-nitrophenoxy, acetic acid, 2-4-nitrophenoxy, acmc-209efx, 4-nitrophenoxy acetic acid
C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)O

Specifications

Undesignated
25g
C8H7NO5
4-nitrophenoxyacetic acid, 2-4-nitrophenoxy acetic acid, 4-nitrophenoxy acetic acid, p-nitrophenoxyacetic acid, acetic acid, 4-nitrophenoxy, 4-nitro-phenoxy-acetic acid, acetic acid, p-nitrophenoxy, acetic acid, 2-4-nitrophenoxy, acmc-209efx, 4-nitrophenoxy acetic acid
C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)O
15720
Crystal-Powder at 20°C
4-Nitrophenoxyacetic Acid
187°C
1798-11-4
MFCD00017030
AVDLFIONKHGQAP-UHFFFAOYSA-N
197.146
197.15
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0781

RTECSNumber : AJ1100000

TSCA : Yes