Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenoxyacetic acid hydrazide, 98%

$64.04 - $727.77

Chemical Identifiers

Chemical Name or Material 4-Nitrophenoxyacetic acid hydrazide
CAS 75129-74-7
Molecular Formula C8H9N3O4
Formula Weight 211.18
MDL Number MFCD00051973
Beilstein 2562952
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1550509

  SDS
alfa aesar™
A1550509
10g Each for $64.04

AAA1550518

  SDS
alfa aesar™
A1550518
50g Each for $244.09

AAA1550530

  SDS
alfa aesar™
A1550530
250g Each for $727.77
Description & Specifications

Specifications

Formula Weight 211.18
Quantity 10g
Health Hazard 1 Warning
Beilstein 2562952
Molecular Formula C8H9N3O4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Nitrophenoxyacetic acid hydrazide
MDL Number MFCD00051973
CAS 75129-74-7
Assay 98%
Melting Point 185° to 192°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.