missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrophenoxyacetic acid hydrazide, 98%

Manufacturer:  Alfa Aesar™ A1550530

 View more versions of this product

Catalog No. AAA1550530



Specifications

Specifications

4-Nitrophenoxyacetic acid hydrazide
250g
C8H9N3O4
2562952
BDPKZWIKLVUGKS-UHFFFAOYSA-N
2-(4-nitrophenoxy)acetohydrazide
156556
98%
185°C to 192°C
75129-74-7
MFCD00051973
2-4-nitrophenoxy acetohydrazide, 4-nitrophenoxyacetic acid hydrazide, acetic acid, 4-nitrophenoxy-, hydrazide, 2-4-nitrophenoxy ethanehydrazide, 4-nitrophenoxy acetic acid hydrazide, acetic acid,2-4-nitrophenoxy-, hydrazide, 2-4-nitro-phenoxy-acetic acid hydrazide
C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)NN
211.177
211.18

Chemical Identifiers

75129-74-7
211.177
BDPKZWIKLVUGKS-UHFFFAOYSA-N
156556
C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)NN
C8H9N3O4
MFCD00051973
2-4-nitrophenoxy acetohydrazide, 4-nitrophenoxyacetic acid hydrazide, acetic acid, 4-nitrophenoxy-, hydrazide, 2-4-nitrophenoxy ethanehydrazide, 4-nitrophenoxy acetic acid hydrazide, acetic acid,2-4-nitrophenoxy-, hydrazide, 2-4-nitro-phenoxy-acetic acid hydrazide
2-(4-nitrophenoxy)acetohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents