Learn More
Alfa Aesar™ 4-Nitrophenoxyacetic acid hydrazide, 98%
Manufacturer: Alfa Aesar™ A1550509
Specifications
4-Nitrophenoxyacetic acid hydrazide | |
10g | |
C8H9N3O4 | |
2562952 | |
BDPKZWIKLVUGKS-UHFFFAOYSA-N | |
2-(4-nitrophenoxy)acetohydrazide | |
156556 | |
98% |
185°C to 192°C | |
75129-74-7 | |
MFCD00051973 | |
2-4-nitrophenoxy acetohydrazide, 4-nitrophenoxyacetic acid hydrazide, acetic acid, 4-nitrophenoxy-, hydrazide, 2-4-nitrophenoxy ethanehydrazide, 4-nitrophenoxy acetic acid hydrazide, acetic acid,2-4-nitrophenoxy-, hydrazide, 2-4-nitro-phenoxy-acetic acid hydrazide | |
C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)NN | |
211.177 | |
211.18 |
Chemical Identifiers
75129-74-7 | |
211.177 | |
BDPKZWIKLVUGKS-UHFFFAOYSA-N | |
156556 | |
C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)NN |
C8H9N3O4 | |
MFCD00051973 | |
2-4-nitrophenoxy acetohydrazide, 4-nitrophenoxyacetic acid hydrazide, acetic acid, 4-nitrophenoxy-, hydrazide, 2-4-nitrophenoxy ethanehydrazide, 4-nitrophenoxy acetic acid hydrazide, acetic acid,2-4-nitrophenoxy-, hydrazide, 2-4-nitro-phenoxy-acetic acid hydrazide | |
2-(4-nitrophenoxy)acetohydrazide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning