missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 157051000

 View more versions of this product

Catalog No. AC157051000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

4-Nitrophenol
112°C to 115°C
Authentic
Plastic bottle
O2NC6H4OH
100 g
01,743; 02,297
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
OC1=CC=C(C=C1)[N+]([O-])=O
139.11
CHEBI:16836
≥98.5% (GC)
100-02-7
Yellow to Brown
99%
C6H5NO3
MFCD00007331
06, 226
15, 6708
BTJIUGUIPKRLHP-UHFFFAOYSA-N
4-nitrophenol
980
139.11
Crystalline Powder, Crystals or Chunks

Chemical Identifiers

100-02-7
139.11
BTJIUGUIPKRLHP-UHFFFAOYSA-N
980
4-nitrophenol
C6H5NO3
MFCD00007331
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
CHEBI:16836
OC1=CC=C(C=C1)[N+]([O-])=O
Safety and Handling

Safety and Handling

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only