Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitronicotinic acid N-oxide, 97%

$86.19 - $293.82

Chemical Identifiers

Chemical Name or Material 4-Nitronicotinic acid N-oxide
CAS 1078-05-3
Molecular Formula C6H4N2O5
Formula Weight 184.11
MDL Number MFCD03426925
Beilstein 176825
Synonym 4-Nitropyridine-3-carboxylic acid N-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2004903

  SDS
alfa aesar™
L2004903
1g Each for $86.19

AAL2004906

  SDS
alfa aesar™
L2004906
5g Each for $293.82
Description & Specifications

Specifications

Formula Weight 184.11
Health Hazard 1 Warning
CAS 1078-05-3
MDL Number MFCD03426925
Assay 97%
Synonym 4-Nitropyridine-3-carboxylic acid N-oxide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 176825
Melting Point ≈170°C
Quantity 1g
Molecular Formula C6H4N2O5
Chemical Name or Material 4-Nitronicotinic acid N-oxide
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.