missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitroindole 98%, ACROS Organics™

Chemical Identifiers

CAS 4769-97-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00010056
Chemical Name or Material 4-Nitroindole, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC124380025 SDS Thermo Fisher Scientific
AC124380025
Glass bottle 2.5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

4769-97-5
100.0
C8H6N2O2
MFCD00010056
≥97.5%
97.5
98%
162.15
4-Nitroindole, 98%

Specifications

Granular Powder
2.5g
203° to 207°C
Glass bottle
Authentic
C8H6N2O2
4769-97-5
≥97.5%
162.15
Yellow
100.0
97.5
MFCD00010056
98%
4-Nitroindole, 98%

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.