missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitroindole-2-carboxylic acid, 97%

$259.38 - $824.11

Chemical Identifiers

CAS 16732-60-8
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
MDL Number MFCD07374290
InChI Key MDWCXENHATUMDQ-UHFFFAOYSA-N
Synonym 1h-indole-2-carboxylic acid, 4-nitro, 4-nitro-1h-indole-2-carboxylic acid, 4-nitro-2-indolecarboxylic acid, 4-nitroindole-2-carboxylic acid
PubChem CID 14368901
SMILES C1=CC2=C(C=C(N2)C(=O)O)C(=C1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63557MD
View Documents
Alfa Aesar™
H63557MD
250mg Each for $259.38
 
AAH6355703
View Documents
Alfa Aesar™
H6355703
1g Each for $824.11
 
Specifications

Chemical Identifiers

16732-60-8
206.157
MDWCXENHATUMDQ-UHFFFAOYSA-N
14368901
C9H6N2O4
MFCD07374290
1h-indole-2-carboxylic acid, 4-nitro, 4-nitro-1h-indole-2-carboxylic acid, 4-nitro-2-indolecarboxylic acid, 4-nitroindole-2-carboxylic acid
C1=CC2=C(C=C(N2)C(=O)O)C(=C1)[N+](=O)[O-]

Specifications

Yellow
16732-60-8
MFCD07374290
MDWCXENHATUMDQ-UHFFFAOYSA-N
206.157
206.15
97%
250mg
C9H6N2O4
1h-indole-2-carboxylic acid, 4-nitro, 4-nitro-1h-indole-2-carboxylic acid, 4-nitro-2-indolecarboxylic acid, 4-nitroindole-2-carboxylic acid
C1=CC2=C(C=C(N2)C(=O)O)C(=C1)[N+](=O)[O-]
14368901
Solid
4-Nitroindole-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures