missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitroindole-2-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H63557MD

 View more versions of this product

Catalog No. AAH63557MD



Specifications

Specifications

4-Nitroindole-2-carboxylic acid
250mg
C9H6N2O4
4-nitro-1h-indole-2-carboxylic acid, 4-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 4-nitro, 4-nitro-2-indolecarboxylic acid
C1=CC2=C(C=C(N2)C(=O)O)C(=C1)[N+](=O)[O-]
14368901
Solid
Yellow
16732-60-8
MFCD07374290
MDWCXENHATUMDQ-UHFFFAOYSA-N
206.157
206.15
97%

Chemical Identifiers

16732-60-8
206.157
MDWCXENHATUMDQ-UHFFFAOYSA-N
14368901
C9H6N2O4
MFCD07374290
4-nitro-1h-indole-2-carboxylic acid, 4-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 4-nitro, 4-nitro-2-indolecarboxylic acid
C1=CC2=C(C=C(N2)C(=O)O)C(=C1)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents