missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitroindole-2-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H6355703

 View more versions of this product

Catalog No. AAH6355703



Specifications

Specifications

4-Nitroindole-2-carboxylic acid
1g
C9H6N2O4
1h-indole-2-carboxylic acid, 4-nitro, 4-nitro-1h-indole-2-carboxylic acid, 4-nitro-2-indolecarboxylic acid, 4-nitroindole-2-carboxylic acid
C1=CC2=C(C=C(N2)C(=O)O)C(=C1)[N+](=O)[O-]
14368901
Solid
Yellow
16732-60-8
MFCD07374290
MDWCXENHATUMDQ-UHFFFAOYSA-N
206.157
206.15
97%

Chemical Identifiers

16732-60-8
206.157
MDWCXENHATUMDQ-UHFFFAOYSA-N
14368901
C9H6N2O4
MFCD07374290
1h-indole-2-carboxylic acid, 4-nitro, 4-nitro-1h-indole-2-carboxylic acid, 4-nitro-2-indolecarboxylic acid, 4-nitroindole-2-carboxylic acid
C1=CC2=C(C=C(N2)C(=O)O)C(=C1)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents