missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitroimidazole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC161605000

 View more versions of this product

Catalog No. AC161605000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Nitroimidazole
Authentic
White to Yellow
500g
97.5
98%
MFCD00005196
1h-imidazole, 4-nitro
VYDWQPKRHOGLPA-UHFFFAOYSA-N
5-nitro-1H-imidazole
18208
113.08
98%
>200°C
Plastic Bottle
303.0°C
3034-38-6
100.0
C3H3N3O2
23, 50
Solubility in water: slightly soluble in water. Other solubilities: soluble in dilute acids,soluble in most common organic solvents
C1=C(NC=N1)[N+](=O)[O-]
113.08
CHEBI:64635
Powder

Chemical Identifiers

3034-38-6
113.08
VYDWQPKRHOGLPA-UHFFFAOYSA-N
18208
5-nitro-1H-imidazole
C3H3N3O2
MFCD00005196
1h-imidazole, 4-nitro
CHEBI:64635
C1=C(NC=N1)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-224-7

SDS Documents