missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitroimidazole, 97%

Manufacturer:  Alfa Aesar™ A1245806

 View more versions of this product

Catalog No. AAA1245806



Specifications

Specifications

4-Nitroimidazole
~303°C (decomposition)
3034-38-6
MFCD00005196
4-nitroimidazole, 4-nitro-1h-imidazole, 1h-imidazole, 4-nitro, 5-nitroimidazole, 4 5-nitroimidazole, imidazole, 4-nitro, 1h-imidazole, 5-nitro, unii-y8u32az5o7, 5 4-nitroimidazole, 4-nitro-3h-imidazole
C1=C(NC=N1)[N+](=O)[O-]
113.076
CHEBI:64635
97%
>200°C (392°F)
5g
C3H3N3O2
2815
VYDWQPKRHOGLPA-UHFFFAOYSA-N
5-nitro-1H-imidazole
18208
113.08

Chemical Identifiers

3034-38-6
113.076
VYDWQPKRHOGLPA-UHFFFAOYSA-N
18208
5-nitro-1H-imidazole
C3H3N3O2
MFCD00005196
4-nitroimidazole, 4-nitro-1h-imidazole, 1h-imidazole, 4-nitro, 5-nitroimidazole, 4 5-nitroimidazole, imidazole, 4-nitro, 1h-imidazole, 5-nitro, unii-y8u32az5o7, 5 4-nitroimidazole, 4-nitro-3h-imidazole
CHEBI:64635
C1=C(NC=N1)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NI7892000

TSCA : TSCA

SDS Documents