Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrohippuric acid, 98%

$50.92 - $227.53

Chemical Identifiers

Chemical Name or Material 4-Nitrohippuric acid
CAS 2645-07-0
Molecular Formula C9H8N2O5
Formula Weight 224.17
MDL Number MFCD00007349
Synonym N-(4-Nitrobenzoyl)glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1234706

  SDS
alfa aesar™
A1234706
5g Each for $50.92

AAA1234714

  SDS
alfa aesar™
A1234714
25g Each for $227.53
Description & Specifications

Specifications

Formula Weight 224.17
Melting Point 128° to 131°C
Molecular Formula C9H8N2O5
MDL Number MFCD00007349
Quantity 5g
Chemical Name or Material 4-Nitrohippuric acid
Assay 98%
Synonym N-(4-Nitrobenzoyl)glycine
CAS 2645-07-0
RTECS Number MC0785000