missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrodiphenylamine, 98+%

Manufacturer:  Alfa Aesar™ L0072806

 View more versions of this product

Catalog No. AAL0072806



Description

Description

Applications
4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

4-Nitrodiphenylamine
211°C (30mmHg)
5g
C12H10N2O2
UN3077
4-ndpa, 4-nitro-n-phenyl-aniline, 4-nitrodifenylamin, 4-nitrodifenylamin czech, 4-nitrodiphenylamine, 4-nitrophenyl phenylamine, benzenamine, 4-nitro-n-phenyl, diphenylamine, 4-nitro, p-nitrodiphenylamine, p-nitrophenylphenylamine
XXYMSQQCBUKFHE-UHFFFAOYSA-N
4-nitro-N-phenylaniline
13271
≥98%
190°C (374°F)
132°C to 135°C
836-30-6
MFCD00007301
2051910
Insoluble in water.
C1=CC=C(C=C1)NC2=CC=C(C=C2)[N+](=O)[O-]
214.224
214.22

Chemical Identifiers

836-30-6
214.224
XXYMSQQCBUKFHE-UHFFFAOYSA-N
13271
C1=CC=C(C=C1)NC2=CC=C(C=C2)[N+](=O)[O-]
C12H10N2O2
MFCD00007301
4-ndpa, 4-nitro-n-phenyl-aniline, 4-nitrodifenylamin, 4-nitrodifenylamin czech, 4-nitrodiphenylamine, 4-nitrophenyl phenylamine, benzenamine, 4-nitro-n-phenyl, diphenylamine, 4-nitro, p-nitrodiphenylamine, p-nitrophenylphenylamine
4-nitro-N-phenylaniline
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 212-646-2

RTECSNumber : JJ9600000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents