missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrocinnamyl alcohol, 98%

$82.04 - $1,343.19

Chemical Identifiers

CAS 1504-63-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00017045
InChI Key LGXXEDSIJZHDBN-OWOJBTEDSA-N
Synonym 4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 4-nitrocinnamyl alcohol, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenol
PubChem CID 5462894
IUPAC Name (E)-3-(4-nitrophenyl)prop-2-en-1-ol
SMILES C1=CC(=CC=C1C=CCO)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1033803 View Documents Alfa Aesar™
A1033803
1g Each for $82.04
 
AAA1033806 View Documents Alfa Aesar™
A1033806
5g Each for $323.85
 
AAA1033814 View Documents Alfa Aesar™
A1033814
25g Each for $1,343.19
 
Description

Description

Applications
4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).

Solubility
Soluble in chloroform, dichloromethane and ethyl Acetate.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1504-63-8
179.175
LGXXEDSIJZHDBN-OWOJBTEDSA-N
5462894
C1=CC(=CC=C1C=CCO)[N+](=O)[O-]
C9H9NO3
MFCD00017045
4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 4-nitrocinnamyl alcohol, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenol
(E)-3-(4-nitrophenyl)prop-2-en-1-ol

Specifications

126°C to 130°C
1504-63-8
MFCD00017045
4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 4-nitrocinnamyl alcohol, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenol
C1=CC(=CC=C1C=CCO)[N+](=O)[O-]
179.175
179.18
4-Nitrocinnamyl alcohol
1g
C9H9NO3
1948238
LGXXEDSIJZHDBN-OWOJBTEDSA-N
(E)-3-(4-nitrophenyl)prop-2-en-1-ol
5462894
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA