missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrochalcone, 99%, ACROS Organics™

Manufacturer:  Acros Organics 169330050

 View more versions of this product

Catalog No. AC169330050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Nitrochalcone
Glass bottle
158°C to 164°C
99%
98.5
99%
O2NC6H4CH=CHCOC6H5
07, 482
WDZGGAFMGIOIQS-DHZHZOJOSA-N
(E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one
5377323
Crystalline Powder
Authentic
Yellow to Brown
5g
1222-98-6
100.0
C15H11NO3
MFCD00007382
4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-]
253.257
253.26
≥98.5%

Chemical Identifiers

1222-98-6
253.257
WDZGGAFMGIOIQS-DHZHZOJOSA-N
5377323
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-]
C15H11NO3
MFCD00007382
4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669
(E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one
Safety and Handling

Safety and Handling

EINECSNumber : 214-949-5

RTECSNumber : FL7075500

TSCA : TSCA

SDS Documents