missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrochalcone, 99%

Manufacturer:  Alfa Aesar™ L1009106

 View more versions of this product

Catalog No. AAL1009106



Specifications

Specifications

4-Nitrochalcone
5g
C15H11NO3
400543
WDZGGAFMGIOIQS-DHZHZOJOSA-N
(E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one
5377323
99%
162°C to 166°C
1222-98-6
MFCD00007382
4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-]
253.257
253.26

Chemical Identifiers

1222-98-6
253.257
WDZGGAFMGIOIQS-DHZHZOJOSA-N
5377323
C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-]
C15H11NO3
MFCD00007382
4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669
(E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FL7075500

TSCA : TSCA

Documents