Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrochalcone, 99%

$127.61 - $127.61

Chemical Identifiers

Chemical Name or Material 4-Nitrochalcone
CAS 1222-98-6
Molecular Formula C15H11NO3
Formula Weight 253.26
MDL Number MFCD00007382
Beilstein 400543
Synonym 2-(4-Nitrobenzylidene)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1009114

 
alfa aesar™
L1009114
25g Each for $127.61
Description & Specifications

Specifications

Formula Weight 253.26
Molecular Formula C15H11NO3
Chemical Name or Material 4-Nitrochalcone
Beilstein 400543
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 99%
MDL Number MFCD00007382
CAS 1222-98-6
Synonym 2-(4-Nitrobenzylidene)acetophenone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 162° to 166°C
Quantity 25g
RTECS Number FL7075500
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.