Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Nitrocatechol sulfate dipotassium salt dihydrate, 97%

$371.00 - $371.00

Chemical Identifiers

Chemical Name or Material 4-Nitrocatechol sulfate dipotassium salt dihydrate
CAS 14528-64-4
Molecular Formula C6H3K2NO7S·2H2O
Formula Weight 347.39 (311.36 Anhydrous)
MDL Number MFCD00149509
Beilstein 3805018
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2332501

 
alfa aesar™
B2332501
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2332506

 
alfa aesar™
B2332506
5g Each for $371.00
Description & Specifications

Specifications

Formula Weight 347.39 (311.36 Anhydrous)
Chemical Name or Material 4-Nitrocatechol sulfate dipotassium salt dihydrate
Assay 97%
Molecular Formula C6H3K2NO7S·2H2O
MDL Number MFCD00149509
CAS 14528-64-4
Beilstein 3805018
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Melting Point >300°C
Quantity 0.25g

Applications
4-Nitrocatechol sulfate dipotassium salt dihydrate is a chromogenic sulfatase substrate. It is used as chemical, organic intermediates.

Solubility
Soluble in water (100 mg/ml).

Notes
Light sensitive. Store away from oxidizing agents, light, heat.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C