Learn More
4-Nitrocatechol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1212222
Description
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
4-Nitrocatechol | |
174°C to 177°C | |
MFCD00007242 | |
1867508 | |
Soluble in water. | |
C1=CC(=C(C=C1[N+](=O)[O-])O)O | |
155.11 | |
CHEBI:16318 | |
≥98% |
3316-09-4 | |
C6H5NO4 | |
100 g | |
4-nitrocatechol, 4-nitropyrocatechol, 1,2-dihydroxy-4-nitrobenzene, 4-nitro-1,2-benzenediol, 3,4-dihydroxy-1-nitrobenzene, unii-sw60ng75en, sw60ng75en, 1,2-benzenediol, 4-nitro, nitrocatechol mix of isomers, 4nc | |
XJNPNXSISMKQEX-UHFFFAOYSA-N | |
4-nitrobenzene-1,2-diol | |
3505109 | |
155.11 |
Chemical Identifiers
3316-09-4 | |
MFCD00007242 | |
CHEBI:16318 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 222-009-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only