Learn More
(4-Nitrobenzyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1528806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(4-Nitrobenzyl)triphenylphosphonium bromide | |
∼280°C (decomposition) | |
MFCD00032108 | |
4-nitrobenzyl triphenylphosphonium bromide, p-nitrobenzyl triphenylphosphonium bromide, phosphonium, p-nitrobenzyl triphenyl-, bromide, 4-nitrobenzyl triphenylphosphoniumbromide, 4-nitrobenzyl triphenyl phosphonium bromide, 4-nitrophenyl methyl triphenylphosphanium bromide, acmc-209gym, phosphonium, 4-nitrophenyl methyl triphenyl-, bromide, wln: wnr d1pr&r&r &e, 4-nitrobenzyltriphenylphosphonium bromide | |
[Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1 | |
478.33 | |
478.33 |
2767-70-6 | |
C25H21BrNO2P | |
5 g | |
IPJPTPFIJLFWLP-UHFFFAOYSA-M | |
(4-nitrophenyl)methyl-triphenylphosphanium;bromide | |
197008 | |
98% |
Chemical Identifiers
2767-70-6 | |
478.33 | |
IPJPTPFIJLFWLP-UHFFFAOYSA-M | |
197008 | |
[Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1 |
C25H21BrNO2P | |
MFCD00032108 | |
4-nitrobenzyl triphenylphosphonium bromide, p-nitrobenzyl triphenylphosphonium bromide, phosphonium, p-nitrobenzyl triphenyl-, bromide, 4-nitrobenzyl triphenylphosphoniumbromide, 4-nitrobenzyl triphenyl phosphonium bromide, 4-nitrophenyl methyl triphenylphosphanium bromide, acmc-209gym, phosphonium, 4-nitrophenyl methyl triphenyl-, bromide, wln: wnr d1pr&r&r &e, 4-nitrobenzyltriphenylphosphonium bromide | |
(4-nitrophenyl)methyl-triphenylphosphanium;bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-444-0
RTECSNumber : TA2390000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only