Please login to your online account to display your discounted pricing

4-Nitrobenzyl triphenylphosphonium bromide, 97%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  338950250

 View more versions of this product

Catalog No. AC338950250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 480.34
Quantity 25g
Color White to light yellow
EINECS Number 220-444-0
RTECS Number TA2390000
Melting Point 270° to 275°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Nitrobenzyl triphenylphosphonium bromide
Assay Percent Range 97%
Molecular Formula C25H23BrNO2P
CAS 2767-70-6
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
MDL Number MFCD00032108

Specifications

Chemical Name or Material 4-Nitrobenzyl triphenylphosphonium bromide
Name Note 97%
CAS 2767-70-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C25H23BrNO2P
Formula Weight 480.34
MDL Number MFCD00032108
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents